Ball Electrode
Supplier: WORLD PRECISION INSTRUMENTS LLC
501284
77662-556EA 40.05 USD
77662-556
Ball Electrode
Electrodes Other Electrodes
The Aaron 940 is designed and engineered to be the most reliable and durable desiccator available today. It is used for electrocauthery, tissue vaporization and Fulguration, homeostasis, tissue debulking, and cosmetic procedure. This ball tip design helps reduce tissue damage and allows for precision tissue manipulation. The ball tip electrode is versatile and can be used for a variety of procedures.
  • For use with economy electrosurgical unit
  • Ball Electrode