Stage Micrometer
Supplier: WORLD PRECISION INSTRUMENTS LLC
500828
77662-380EA 304.91 USD
77662-380
Stage Micrometer
Micrometers
  • Suitable for upright microscopes at 40X, 100X and 400X
  • Marks at 50 and 100 μm
  • Scale range: Linear 1 mm/100/0.01 mm
  • 1×3" slide with 1 mm linear target etched into the glass down to 10 μm marks
  • Micrometer type: Line