Scissors, Biemer
Supplier: WORLD PRECISION INSTRUMENTS LLC
500381
77662-340EA 161.69 USD
77662-340
Scissors, Biemer
Scissors
Cuts vessels perpendicularly and without slipping.
  • Spring handle
  • Straight
  • Stainless steel