USB Thumb Drive
Supplier: WORLD PRECISION INSTRUMENTS LLC
300749
77662-074EA 100.11 USD
77662-074
USB Thumb Drive
USB Flash Drives
This USB thumb drive is loaded with the Python digital output package for use with the popular EVOM™ Manual TEER (transepithelial electrical resistance) meter.
A blank replacement thumb drive is also available. This thumb drive hold 32 Gb of data and is suitable for use with the EVOM™ Manual for auto data logging of all your measurements. The retractable design of this SanDisk thumb drive protects the USB connector when the unit is not in use.