Glass Beads for Dry Sterilizer
Supplier: WORLD PRECISION INSTRUMENTS LLC
14405
77661-902EA 86.24 USD
77661-902
Glass Beads for Dry Sterilizer
Beads Glass Beads
  • Replacement glass beads for germinator 500 sterilizer
  • Lead free 1.5 mm diameter glass beads