Scalpel Handles for Disposable Blade
Supplier: WORLD PRECISION INSTRUMENTS LLC
14134-G 14134
77661-796EA 342 USD
77661-796 77661-794
Scalpel Handles for Disposable Blade
Scalpels and Scalpel Assemblies Scalpel Handles
This reusable surgical instrument lets you securely hold and manipulate disposable scalpel blades during surgical procedures. The unique concave/convex jaw prevents the blades from slipping, and the locking mechanism on the long handle secures the blade in place.
  • Convex/concave jaws
  • Round tips
  • Special jaws prevent blades from slipping
The scissor-like handles give excellent control and leverage for the user. This scalpel handle may be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. This scalpel handle is available in surgical grade stainless steel or our premium German steel.