Stevenson Retractor
Supplier: WORLD PRECISION INSTRUMENTS LLC
14131
77661-786EA 98.57 USD
77661-786
Stevenson Retractor
Retractors
  • 3×3 Prongs
  • 3 mm Deep
  • Maximum spread of 15 mm
  • Stainless steel grade : Stevens retractor, standard