Round Handled Needle Holder, Straight
Supplier: WORLD PRECISION INSTRUMENTS LLC
14080
77661-704EA 181.71 USD
77661-704
Round Handled Needle Holder, Straight
Dissecting and Inoculating Loop and Needle Holders
These round-handled needle holders are a popular choice for researchers doing small animal surgeries. The knurled handle provides a secure grip on the surgical instrument and reduces the chance of it slipping during use. The round handle is also more comfortable, especially for long procedures.
  • Fine (0.3 mm) smooth tips
  • Stainless steel
  • Straight smooth jaws
The round handles allow you to roll the instrument between your fingers as you are working, giving you more precise control over the maneuvering of the tip holding the surgical needle. The knurled handle lets you apply the correct amount of pressure for accurate suturing. The smooth jaw of the needle holder reduces the amount of trauma and tearing of delicate tissues and improves the visibility of the working area during a procedure.