Luer Valve Assortment Kit
Supplier: WORLD PRECISION INSTRUMENTS LLC
14011
77661-658EA 530 USD
77661-658
Luer Valve Assortment Kit
Valves Injection Valves
This Luer fittings kit is useful for building your own liquid flow experiment. It provides the means to start, stop, add, divide and control a flow of liquid or gas.
  • Over 300 assorted Luer fittings and parts
  • Luer fittings for quick and easy connect and disconnect
  • Applications: Liquid flow experimental setups
Included in the kit are over 300 assorted parts such as one-way and 3-way stopcocks, manifolds, Y-connectors, injection sites, male and female luer caps, check valves, syringe-activated check valves, slide clamps, roller clamps, and pinch clamps. All (except clamps) have a luer fitting for quick and easy connecting and disconnecting. This Luer fittings kit ncludes assorted luer fittings for use with flexible tubing.