VWR® Ultra Low Temperature Freezers, with 4 Inner Doors, –40 °C, 230 V, 50 Hz
Supplier: VWR International
76307-988 76307-994 77661-486
76307-988EA 13632.32 USD
76307-988 76307-994 77661-486
VWR® Ultra Low Temperature Freezers, with 4 Inner Doors, –40 °C, 230 V, 50 Hz
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by green, hydrocarbon refrigerants delivering operational savings and sustainability.
  • 230 V models
  • Low energy
  • Low peak variation
  • Fast door opening recovery
  • Long warm up time
  • Three sizes
VWR -40 °C ULT freezers are built for seamless operation with user-friendly features. With low noise operation, flexible rack configurations and faster re-entry times, these -40 °C ULT freezers are designed to enhance the overall user experience and meet the needs of busy laboratories.
Certifications:
cULus, SNAP
Ordering information:
Options and accessories are sold separately; contact your VWR representative for more information.
Caution:
230 V models. Corresponding models with voltage suitable for use in US (76307-984, 76307-990 and 76307-482) are also available, please see separate product page.
Must be removed from pallet upon delivery. Fits through standard 34.5" doorways.