Notrax® 525G Sani-Flex™ Anti-fatigue and Safety Matting
Supplier: SUPERIOR MANUFACTURING GROUP INC
526S0023GY
77661-456EA 81.6 USD
77661-456
Notrax® 525G Sani-Flex™ Anti-fatigue and Safety Matting
Floor Mats Traction and Anti-Fatigue Floor Mats
Closed anti-slip gritted structure surface. Superb anti-fatigue properties due to its unique design underneath.
  • Closed anti-slip gritted structure surface
  • Superb anti-fatigue properties due to its unique design underneath
  • Heavy duty 100% nitrile rubber compound, designed to yield a long service life
Specially designed for the lab industry, Sani-Flex™ anti-fatigue matting provides the optimum combination of comfort, anti-slip surface, quality and durability, as well as oil/grease and chemical resistance. Its smooth surface and rounded off edges make it easy to clean. Made of tough rubber compounds, designed to yield a long service life. Aggressive slip-resistant surface.