Clean-Step™ Adhesive Peel Off Surface Tacky Mats
Supplier: NOTRAX USA, INC.
080S263800 080S284700 080S203800 080S204700
77661-412EA 152.38 USD
77661-412 77661-420 77661-404 77661-406
Clean-Step™ Adhesive Peel Off Surface Tacky Mats
Floor Mats Tacky Mats
Clean-Step™ tacky mats traps and keeps out impurities, dust and particles. Numbered tabs ensure one sheet at a time removal and indicate number of remaining layers. The sticky mat is not damaged by footwear or wheels, does not slip, is noncontaminating. Blue and grey are B level tack whereas white is A level, high level of tack.
  • Helps prevent patient to patient and healthcare worker to patient cross contamination and infection
  • Remove and contain dirt and dust from foot traffic and equipment wheels
  • Traps and keeps out impurities, dust, and particulates
  • Adhesive surface grabs and holds soil matter with no transfer
  • Peel away sheets for fresh, clean surface
  • Anti-microbial
  • Low profile numbered tabs
Type: Ergonomic, anti-fatigue and safety matting
Environment: Dry areas