Notrax® 080 CleanStep™ Adhesive Peel Off Surface Tacky Mats
Supplier: NOTRAX USA, INC.
080S2645BU 080S2645GY 080S2645WH 080S1836BU 080S1836GY 080S1836WH 080S1845BU 080S1845GY 080S1845WH 080S2436BU 080S2436GY
77661-414CS 128.9 USD
77661-414 77661-416 77661-418 77661-392 77661-394 77661-396 77661-398 77661-400 77661-402 77661-408 77661-410
Notrax® 080 CleanStep™ Adhesive Peel Off Surface Tacky Mats
Floor Mats Tacky Mats
Clean-Step™ tacky mats traps and keeps out impurities, dust and particles. Numbered tabs ensure one sheet at a time removal and indicate number of remaining layers.
  • Helps prevent patient to patient and healthcare worker to patient cross contamination and infection
  • Remove and contain dirt and dust from foot traffic and equipment wheels
  • Adhesive surface grabs and holds soil matter with no transfer
  • Peel away sheets for fresh, clean surface
  • Anti-microbial
The sticky mat is not damaged by footwear or wheels, does not slip, is noncontaminating. Blue and gray are B level tack whereas white is A level, high level of tack.
Group: Ergonomic, anti-fatigue and safety matting
Environment: Dry areas
Usage intensity: Medium traffic