AlerTox® Milk Lateral Flow Test Sticks
Supplier: HARDY DIAGNOSTICS
KIT3015
77661-324PK 423.26 USD
77661-324
AlerTox® Milk Lateral Flow Test Sticks
Food and Beverage Test Systems
Designed specifically to help manufacturers keep items free of antigens that may cause allergic reactions.
  • Quick and reliable
  • No cross-reactivity
  • Results in 10 minutes
  • High sensitivity
AlerTox® Sticks are a rapid, user-friendly test for the detection and qualification of main food allergens with no lab equipment or scientific knowledge required.