4'-Methoxybutyrophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-Methoxyphenyl)butan-1-one

47676-25G
77659-624EA 59 USD
77659-624
4'-Methoxybutyrophenone ≥97%
4'-Methoxybutyrophenone

Caution: For Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00027138
CAS Number: 4160-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR