1-Phenyl-1-hexyn-3-ol ≥97%

Supplier: AOBChem USA

Warning

258768-5G 258768-25G
77634-516EA 66 USD
77634-516 77654-484
1-Phenyl-1-hexyn-3-ol ≥97%
1-Phenyl-1-hexyn-3-ol

Caution: For Research Use Only

Formula: C₁₂H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00046687
CAS Number: 1817-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR