Dicarbonylcyclopentadienylcobalt 95% (by titrimetric analysis)

Supplier: Ambeed
Danger

Synonyms: Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)

A915754-250MG A915754-5G
77603-292EA 20.5 USD
77603-292 77627-752
Dicarbonylcyclopentadienylcobalt 95% (by titrimetric analysis)
Dicarbonylcyclopentadienylcobalt

Caution: for Research Use Only

Formula: C₇H₅CoO₂
MDL Number: MFCD00009589
CAS Number: 12078-25-0
UN: 1992
ADR: 3,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR