5-Chloro-2-hydroxy-3-methylbenzaldehyde ≥97%

Supplier: Ambeed
Warning

A759601-1G
77627-512EA 54.59 USD
77627-512
5-Chloro-2-hydroxy-3-methylbenzaldehyde ≥97%
5-Chloro-2-hydroxy-3-methylbenzaldehyde

Caution: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.60 g/mol
MDL Number: MFCD00275687
CAS Number: 23602-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR