Anisomycin ≥98%

Supplier: Ambeed
Warning

Synonyms: (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate

A646979-10MG A646979-1MG A646979-50MG A646979-100MG A646979-250MG
77627-290EA 42.23 USD
77627-290 77627-292 77627-294 77878-466 77878-467
Anisomycin ≥98%
Anisomycin

Caution: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD00077650
CAS Number: 22862-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR