Pentoxifylline ≥99%

Supplier: Ambeed
Warning

Synonyms: 3,7-Dimethyl-1-(5-oxohexyl)xanthine, 1-(5-Oxohexyl)theobromine, 3,7-Dimethyl-1-(5-oxohexyl)-1H-purine-2,6(3H,7H)-dione, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione

A646154-1MG
77627-288EA 6.18 USD
77627-288
Pentoxifylline ≥99%
Pentoxifylline

Caution: for Research Use Only

Formula: C₁₃H₁₈N₄O₃
MW: 278.31 g/mol
MDL Number: MFCD00063379
CAS Number: 6493-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR