3,5-Dichloro-4-hydroxybenzonitrile ≥98%

Supplier: Ambeed
Danger

Synonyms: Chloroxynil

A587627-5G
77627-220EA 142.14 USD
77627-220
3,5-Dichloro-4-hydroxybenzonitrile ≥98%
3,5-Dichloro-4-hydroxybenzonitrile

Caution: for Research Use Only

Formula: C₇H₃Cl₂NO
MDL Number: MFCD00002177
CAS Number: 1891-95-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR