(1-Butyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: Butyltriphenylphosphonium bromide

A427714-1000G A427714-100G A427714-500G A427714-10G A427714-25G
76695-036EA 59 USD
76695-036 76695-038 76695-040 77601-836 77626-854
(1-Butyl)triphenylphosphonium bromide 97%
(1-Butyl)triphenylphosphonium bromide

Caution: for Research Use Only

Formula: H₃C(CH₂)₃P(C₆H₅)₃Br
MDL Number: MFCD00011855
CAS Number: 1779-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR