7-Hydroxycoumarin-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A358382-100MG A358382-1G A358382-250MG A358382-5G A358382-25G
76694-182EA 12 USD
76694-182 76694-184 76694-186 77601-588 77626-694
7-Hydroxycoumarin-3-carboxylic acid 98%
7-Hydroxycoumarin-3-carboxylic acid

Caution: for Research Use Only

Formula: C₁₀H₆O₅
MDL Number: MFCD00017491
CAS Number: 779-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR