Copper(II) sulfide ≥99%

Supplier: Ambeed
Danger

Synonyms: Copper sulfide, Cupric sulfide, Copper sulphide, Cupric sulphide, Copper(II)sulfide, Copper(II)sulphide

A355482-100G
76694-138EA 6.37 USD
76694-138 77626-686
Copper(II) sulfide ≥99%
Copper (II) sulphide

Caution: for Research Use Only

Formula: CuS
MDL Number: MFCD00016066
CAS Number: 1317-40-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR