Cholesterol acetate ≥95%

Supplier: Ambeed
Warning

Synonyms: (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl acetate, Cholesteryl Acetate, Acetyl Cholesterol, Acetic Acid Cholesterol Ester

A350738-25G A350738-100G
77626-670EA 90.64 USD
77626-670 77879-757
Cholesterol acetate ≥95%
Cholesterol acetate

Caution: for Research Use Only

Formula: C₂₉H₄₈O₂
MW: 428.70 g/mol
MDL Number: MFCD00003636
CAS Number: 604-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR