Methyl N-acetyl-α-D-glucosaminide ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-acetamido-2-deoxy-alpha-d-glucopyranoside, N-((2S,3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3-yl)acetamide, Methyl 2-acetamido-2-deoxy-α-D-glucopyranoside
A329314-1G
A329314-250MG
A329314-5G
77626-626EA
135.96
USD
77626-626
77626-628
77878-035
Methyl N-acetyl-α-D-glucosaminide ≥95%
Methyl N-acetyl-α-D-glucosaminide
Caution: for Research Use Only
Formula:
C₉H₁₇NO₆ |
MDL Number:
MFCD00056076 CAS Number: 6082-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...