D-(+)-Glucose monohydrate 98+%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

A297831-500G
76693-348EA 11 USD
76693-348 77626-520
D-(+)-Glucose monohydrate 98+%
D(+)-Glucose monohydrate

Caution: for Research Use Only

Formula: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
Melting Pt: 83
MDL Number: MFCD00149450
CAS Number: 14431-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR