2',4',5'-Trifluoroacetophenone ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone

A230504-5G
77626-212EA 67.98 USD
77626-212
2',4',5'-Trifluoroacetophenone ≥98%
2',4',5'-Trifluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₅F₃O
MDL Number: MFCD00061193
CAS Number: 129322-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR