4-Hydroxy-2-quinolinecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyquinoline-2-carboxylic Acid, Transtorine, Empirical Formula, Kynurenic acid, Quinurenic acid, KYNA, Kynuronic acid, Kinurenic acid

A141971-100MG A141971-250MG A141971-25MG A141971-25G A141971-5G
103803-084EA 32 USD
103803-084 76798-346 76798-348 76798-350 77316-286 77624-176
4-Hydroxy-2-quinolinecarboxylic acid 97%
4-Hydroxy-2-quinolinecarboxylic acid

Caution: for Research Use Only

Formula: C₁₀H₇NO₃
Boiling Pt: 358.4 °C (760 mm Hg)
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD00149476
CAS Number: 492-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR