Diethyl pyrocarbonate 97%

Supplier: Ambeed
Warning

A127898-10G A127898-1G A127898-25G A127898-5G A127898-100G A127898-500G
103831-908EA 8 USD
103831-908 76797-756 76797-758 77298-252 77298-254 77599-860 77624-072
Diethyl pyrocarbonate 97%
Diethyl pyrocarbonate

Caution: for Research Use Only

Formula: O(COOC₂H₅)₂
Boiling Pt: 93…94 °C at 18 mmHg
Flash Pt: 61.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR