tri-Sodium citrate dihydrate ≥99%

Supplier: Ambeed
Warning

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

A122812-1000G A122812-500G A122812-100G
77596-470EA 14.35 USD
77596-470 77596-472 77624-020
tri-Sodium citrate dihydrate ≥99%
tri-Sodium citrate dihydrate

Caution: for Research Use Only

Formula: Na₃C₆H₅O₇·2H₂O
MDL Number: MFCD00150031
CAS Number: 6132-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR