Handheld UV Lamp, 365 nm
Supplier: Hardy Diagnostics
LSS3
77623-674EA 143.71 USD
77623-674
Handheld UV Lamp, 365 nm
Lights UV Lights, Lamps and Bulbs
UV lamp, battery operated, hand-held lamp.
  • High intensity
  • 365 nm wavelength
  • 3 LED lights
  • rechargeable battery
Includes charger and USB cable, 6" long, ideal for detecting fluorescent colonies on a Petri plate.