GlutenTox® Home Gluten Detection Kit
Supplier: HARDY DIAGNOSTICS
KIT3009
77623-672PK 84.4 USD
77623-672
GlutenTox® Home Gluten Detection Kit
Food and Beverage Test Systems
GlutenTox®, Home is a friendly gluten detection kit for foods and drinks. GlutenTox Home contains the G12 anti-gliadin antibody that specifically recognizes the gluten fraction that triggers celiac disease.
  • Detects gluten from wheat, barley, rye and rare varieties of oat
  • Adjustable sensitivity for 5 or 20 ppm of gluten
  • Results in less than 20 minutes
  • No special equipment required
  • Safe and easy to use