Carbol fuchsin (Ziehl-Neelsen)

Supplier: HARDY DIAGNOSTICS

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

484A32
77623-666EA 145.05 USD
77623-666
Carbol fuchsin (Ziehl-Neelsen)
Carbol fuchsin

An aqueous solution of basic fuchsin, phenol and reagent alcohol. 'Hot method', requires heating of the slide.

Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
MDL Number: MFCD00013129
CAS Number: 4197-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR