Swab, Foam Tip, Sterile
Supplier: Hardy Diagnostics
2515061PFB
77623-664PK 577.09 USD
77623-664
Swab, Foam Tip, Sterile
Swabs Swabs for Sampling
The Puritan 6" sterile standard foam swab is perfect for collecting nasal and anterior nares specimens for diagnostic testing, buccal and other cell collection for testing and screening.
  • Tip material: Foam
  • Tip type: Polystyrene
DNA testing and diagnostics cell collection. The swab has a high particle collection capacity and is individually wrapped, presented in 50 perforated strips of 20, and remains sterile until you are ready to use and easy to store. Made in the USA.