SHOWA CN750 Chemical Resistant Gloves
Supplier: Showa
CN750L-09 CN750M-08 CN750S-07 CN750XL-10 CN750XXL-11
77623-516BG 57.06 USD
77623-516 77623-518 77623-520 77623-522 77623-524
SHOWA CN750 Chemical Resistant Gloves
Gloves
The SHOWA CN750 features a high level of chemical resistance. At 320 mm in length, the glove is designed with a grip wave pattern to ensure a secure grip during use, even in challenging conditions.
  • Oil-, chemical-resistant
  • Forearm protection
  • Impermeable and durable
  • Easy donning and doffing
  • Eco Best Technology® (EBT)
  • Unlined and wave pattern finish
The interior of the glove incorporates innovative technology. The flocking particles help to distribute moisture, making it easier to put on and take off the gloves even after extended wear. The SHOWA CN750 also features Eco Best Technology, making it a biodegradable glove.
Applications:
- Chemical spray and treatment
- Handling light chemicals in agriculture, horticulture
- Light assembly of oil-coated pieces
- Coating preparation
- Washing and cleaning
Certifications: CE cat. III, EN 388:2016+A1:2018, EN ISO 374-1:2016+A1:2018/Type, EN ISO 374-5:2016, EN 420:2003+A1:2009, Food contact certified.
Packaging: Pair per polybag: 12, polybags per case: 6, pair per case: 72.