Chemical Resistant Gloves, 732
Supplier: Showa
732-07 732-08 732-09 732-10 732-11
77623-486CS 276.77 USD
77623-486 77623-488 77623-490 77623-492 77623-494
Chemical Resistant Gloves, 732
Gloves
The 100% nitrile glove provides protection against a broad range of oils, solvents and chemicals. Diamond grip surface finish increases wet grip for enhanced ease and safety. A flocked liner makes the SHOWA 732 easy to remove.
  • Forearm protection
  • Chemical-resistant; Oil-resistant
  • Impermeable
  • Textured finish
  • Ergonomic
Trades and applications:
-Poultry, meat and seafood processing
-Light chemical handling in horticulture, agriculture
-Light assembly of oil-coated pieces
-Painting and spray workshops
-Chemical spray and treatment
-Washing and cleaning
-Food packing and handling
Certifications: CE Cat III, EN 388:2016+A1:201, EN ISO 374-1:2016+A1:2018/Type, EN ISO 374-5:2016, SO 18889, EN ISO 21420:2020, Food contact.
Packaging: 12 pair per polybag, 12 polybags per case, 144 pair per case.