Quercetin hydrate ≥95%

Supplier: Ambeed
Danger

Synonyms: 3,3',4',5,7-Pentahydroxyflavone hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one xhydrate

A1257621-25G
77621-312EA 23.69 USD
77621-312
Quercetin hydrate ≥95%
Quercetin hydrate

Caution: for Research Use Only

Formula: C₁₅H₁₀O₇·xH₂O
MW: 302.24 g/mol (anhy)
MDL Number: MFCD00006828
CAS Number: 849061-97-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR