Crystal violet BS

Supplier: Ambeed
Danger

A1003341-500G A1003341-1000G
76646-116EA 8.49 USD
76646-116 77595-648 77619-502
Crystal violet BS
Crystal Violet

Caution: for Research Use Only

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR