VWR® 550 Benchtop Conductivity Meters
Supplier: VWR International
VWR-EC500
77618-948EA 539.98 USD
77618-948
VWR® 550 Benchtop Conductivity Meters
Electrochemistry Meters Conductivity Meters
The VWR-EC550 meter provide accurate and reliable measurements of conductivty, TDS (total dissolved solids), and temperature in any environment.
  • High resolution LCD display (6.0")
  • 1 point calibration with standard recognition
  • IP54 protection
  • Automatic or manual temperature compensation capabilities
  • Auto-read and continuous-read measurement options
  • Ability to save 50 data sets
  • Ability to set cell constant value
  • Three-year meter warranty
The easy to use VWR-EC550 benchtop meters provide accurate conductivity measurements within any laboratory environment. Easily choose a kit to fit your application needs by selecting the required cell constant for you sample conductance range.
Ordering information: For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the related information section.
Delivery information: All meters include electrode stand and silicone in-use cover.