VWR® 700 Portable Dissolved Oxygen Meters
Supplier: VWR International
VWR-DO700A
77618-930EA 941.97 USD
77618-930
VWR® 700 Portable Dissolved Oxygen Meters
Electrochemistry Meters Oxygen Meters
The VWR-DO700 meter provide accurate and reliable measurements of dissolved oxygen concentration and dissolved oxygen saturation.
  • High resolution LCD display (3.5")
  • Air-saturated water or zero oxygen calibration capabilities
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Data capacity of 500 sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Auto barometric pressure compensation
The easy to use VWR-DO700 portable meters provide accurate dissolved oxygen measurements for accurate water quality testing. The large, backlit LCD display allows results to easily be viewed during operation. Easily transport the meter and accessories with the included carrying case.
Three-year meter warranty.
Delivery information: All meters include electrode holders (2), meter silicone rubber case, carrying case and hand strap.