VWR® Automatic Temperature Compensation (ATC) Probe, Avantor
Supplier: VWR International
TEMP01
77618-918EA 102.18 USD
77618-918
VWR® Automatic Temperature Compensation (ATC) Probe, Avantor
Temperature Probes
For connection to VWR® Electrochemistry meters to allow for Automatic Temperature Compensation.
  • Sensor material: 316 stainless steel
  • Temperature compensation unit: 2.25 KΩ