VWR® ORP Electrode, Avantor
Supplier: VWR International
77618-866
77618-866EA 146.5 USD
77618-866
VWR® ORP Electrode, Avantor
Electrodes Redox Electrodes
The ORP01 Electrode provides the oxidation-reduction potential of a sample (dependent on meters capabilities).
  • Sensor material: Glass
  • Reference type: Ag/AgCl