Dehydrocholic acid ≥95%

Supplier: Ambeed
Warning

A956430-1G A956430-5G A956430-100G
77618-636EA 6.18 USD
77618-636 77618-638 77749-787
Dehydrocholic acid ≥95%
Dehydrocholic acid

Caution: for Research Use Only

Formula: C₂₄H₃₄O₅
MDL Number: MFCD00066410
CAS Number: 81-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR