2-Aminoacridone ≥95%

Supplier: Ambeed
Warning

A674902-5MG
77617-814EA 50.29 USD
77617-814
2-Aminoacridone ≥95%
2-Aminoacridone

Caution: for Research Use Only

Formula: C₁₃H₁₀N₂O
MDL Number: MFCD00037397
CAS Number: 27918-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR