Genkwanin, 7-methoxyapigenin ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-(4-hydroxyphenyl)-7-methoxy-4H-chromen-4-one, 4',5-Dihydroxy-7-methoxyflavone

A400206-10MG A400206-1MG A400206-5MG
77616-726EA 32.96 USD
77616-726 77616-728 77616-730
Genkwanin, 7-methoxyapigenin ≥98%
Genkwanin, 7-methoxyapigenin

Caution: for Research Use Only

Formula: C₁₆H₁₂O₅
MW: 284.27 g/mol
MDL Number: MFCD00017452
CAS Number: 437-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR