2,5-Dimercapto-1,3,4-thiadiazole ≥98%

Supplier: Ambeed
Danger

Synonyms: 1,3,4-Thiadiazole-2,5-dithiol

A305906-100G A305906-500G A305906-25G
77616-318EA 43.26 USD
77616-318 77616-320 77685-872
2,5-Dimercapto-1,3,4-thiadiazole ≥98%
2,5-Dimercapto-1,3,4-thiadiazole

Caution: for Research Use Only

Formula: C₂H₂N₂S₃
MW: 150.25 g/mol
MDL Number: MFCD00003103
CAS Number: 1072-71-5
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR