1-Bromo-3,5-diiodobenzene 95%

Supplier: Ambeed
Warning

A210168-1G A210168-250MG A210168-25G A210168-5G A210168-10G
77237-370EA 8 USD
77237-370 77237-372 77237-374 77237-376 77615-774
1-Bromo-3,5-diiodobenzene 95%
1-Bromo-3,5-diiodobenzene

Caution: for Research Use Only

Formula: C₆H₃BrI₂
MDL Number: MFCD07778997
CAS Number: 149428-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR