6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%

Supplier: Ambeed
Warning

A123399-10G A123399-25G A123399-5G A123399-250MG
103800-400EA 9 USD
103800-400 103831-236 77284-970 77284-972 77312-520 77614-970
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione

Caution: for Research Use Only

Formula: C₃H₂BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00154757
CAS Number: 4956-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR