Pantoprazole sulfide 98%

Supplier: Ambeed
Warning

Synonyms: 5-Difluoromethoxy-2-[(3,4-dimethoxy-2-pyridyl)methyl]thio-1H-benzimidazole, 5-(Difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)thio)-1H-benzo[d]imidazole, 5-(Difluoromethoxy)-2-[[(3,4-dimethoxy-2-pyridyl)methyl]thio]benzimidazole

A100634-10G A100634-25G A100634-100G
103821-240EA 10 USD
103821-240 77270-164 77270-166 77614-728
Pantoprazole sulfide 98%
Pantoprazole sulfide

Caution: for Research Use Only

Formula: C₁₆H₁₅F₂N₃O₃S
MW: 367.37 g/mol
Boiling Pt: 526.3 °C (760 mm Hg)
Melting Pt: 110…118 °C
Flash Pt: 272.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368273
CAS Number: 102625-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR